Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012221.1 | NP_001012221.1 | 1 | 1386 | ||
GenBank mRNA | BC083681.1 | - | 1 | 1386 | 7135892 |
Gene Symbol: LOC100361036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764112.1 | XP_008762334.1 | 1 | 1936 |