Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238946.2 | XP_006239008.1 | 6 | 819 | ||
XM_006238947.2 | XP_006239009.1 | 6 | 800 | |||
XM_006238948.2 | XP_006239010.1 | 6 | 819 | |||
XM_006238950.2 | XP_006239012.1 | 6 | 819 | |||
XM_006238951.2 | XP_006239013.1 | 6 | 800 | |||
XM_006238953.2 | XP_006239015.1 | 6 | 820 | |||
XM_008764158.1 | XP_008762380.1 | 7 | 921 |