Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135803.1 | NP_001129275.1 | 6 | 824 | ||
XM_006239990.2 | XP_006240052.1 | 6 | 755 | |||
XM_006239991.1 | XP_006240053.1 | 7 | 982 | |||
XM_006239992.2 | XP_006240054.1 | 7 | 1259 | |||
XM_006239993.2 | XP_006240055.1 | 5 | 668 | |||
XM_008764601.1 | XP_008762823.1 | 5 | 630 |