Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764911.1 | XP_008763133.1 | 106, 107 | 19773 | ||
GenBank mRNA | BC098809.1 | - | 4, 5 | 470 |