Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297609.1 | NP_001284538.1 | 8 | 1637 | ||
NM_004672.4 | NP_004663.3 | 9 | 1661 | |||
XM_017002771.1 | XP_016858260.1 | 7 | 1042 | |||
XM_017002772.1 | XP_016858261.1 | 9 | 2832 | |||
GenBank mRNA | AB167411.1 | - | 9 | 1392 | ||
AB208805.1 | - | 8 | 1106 | |||
AF100318.1 | - | 6 | 806 | |||
BC015914.1 | - | 5 | 563 | |||
BC129950.1 | - | 1 | 91 | |||
BC129951.1 | - | 1 | 91 |