Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000592.1 | NP_001000592.1 | 1 | 250 |
Gene Symbol: Olr1082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173335.1 | NP_775457.1 | 1 | 250 | ||
GenBank mRNA | M64381.1 | - | 1 | 250 |