Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185072.2 | NP_001172001.1 | 4 | 509 | ||
NM_001185073.2 | NP_001172002.1 | 3 | 470 | |||
NM_012129.4 | NP_036261.1 | 3 | 466 | |||
GenBank mRNA | AK312480.1 | - | 4 | 394 | ||
AL136770.1 | - | 3 | 318 | |||
BC036754.2 | - | 2 | 255 | 5201423 | ||
BC068532.1 | - | 3 | 352 | 5294796 | ||
DA121583.1 | - | 3 | 313 | |||
DA230901.1 | - | 3 | 407 | |||
DA330572.1 | - | 4 | 394 | |||
DB062973.1 | - | 3 | 468 | |||
DB224591.1 | - | 3 | 356 |