Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASH2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105214.2 | NP_001098684.1 | 12 | 1512 | ||
NM_001261832.1 | NP_001248761.1 | 12 | 1512 | |||
NM_001282272.1 | NP_001269201.1 | 11 | 1265 | |||
NM_004674.4 | NP_004665.2 | 12 | 1411 | |||
XM_005273682.1 | XP_005273739.1 | 13 | 1957 | |||
XM_005273683.1 | XP_005273740.1 | 13 | 1530 | |||
XM_006716412.1 | XP_006716475.1 | 13 | 1957 | |||
XM_006716413.2 | XP_006716476.1 | 12 | 1377 | |||
GenBank mRNA | AB020982.1 | - | 12 | 1364 | ||
AF056717.1 | - | 12 | 1366 | |||
AF056718.1 | - | 12 | 1357 | |||
AK291750.1 | - | 12 | 1375 | |||
AK291938.1 | - | 12 | 1383 | |||
AK293699.1 | - | 5 | 489 | |||
AK298483.1 | - | 11 | 1208 | |||
BC015936.2 | - | 12 | 1379 | 3921999 |