Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108992.1 | NP_001102462.1 | 10 | 1343 | ||
XM_006242230.1 | XP_006242292.1 | 10 | 1340 | |||
XM_006242231.1 | XP_006242293.1 | 9 | 1283 | |||
XM_006242233.1 | XP_006242295.1 | 9 | 1136 | |||
XM_006242234.1 | XP_006242296.1 | 8 | 1079 | |||
XM_006242235.1 | XP_006242297.1 | 9 | 923 | |||
XM_006242236.2 | XP_006242298.1 | 10 | 1107 | |||
GenBank mRNA | AY724510.1 | - | 8 | 1019 | ||
BC166484.1 | - | 8 | 872 | |||
FQ211850.1 | - | 4 | 427 |