Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160100.1 | NP_001153572.1 | 4 | 725 | ||
NM_006984.4 | NP_008915.1 | 4 | 614 | |||
NM_182848.3 | NP_878268.1 | 4 | 782 | |||
XM_011521134.2 | XP_011519436.1 | 5 | 707 | |||
XM_017020843.1 | XP_016876332.1 | 5 | 919 | |||
XM_017020844.1 | XP_016876333.1 | 5 | 882 | |||
GenBank mRNA | AF339794.1 | - | 1 | 50 | ||
AK055855.1 | - | 4 | 777 | |||
AK315737.1 | - | 4 | 566 | |||
BC010920.1 | - | 4 | 586 | 4246806 | ||
CR456845.1 | - | 4 | 553 | |||
U89916.1 | - | 4 | 577 |