Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108995.2 | NP_001102465.2 | 10 | 1025 | ||
XM_006242652.2 | XP_006242714.1 | 10 | 1306 | |||
XM_006242653.2 | XP_006242715.1 | 9 | 1061 | |||
GenBank mRNA | BC166711.1 | - | 10 | 1025 | 7930337 |