Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013248.1 | NP_001013266.1 | 2 | 1433 | ||
GenBank mRNA | BC089975.1 | - | 2 | 1433 | 7316995 |