Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171092.1 | NP_001164563.1 | 2 | 970 | ||
NM_001171095.1 | NP_001164566.1 | 2 | 739 | |||
NM_020384.3 | NP_065117.1 | 2 | 805 | |||
GenBank mRNA | AF177340.1 | - | 2 | 970 | ||
AF250558.1 | - | 1 | 451 | |||
AK075371.1 | - | 2 | 681 | |||
AK075405.1 | - | 2 | 777 | |||
AK312515.1 | - | 2 | 739 | |||
AY358474.1 | - | 1 | 572 | |||
BC014424.1 | - | 1 | 572 | 4645075 | ||
BC071747.1 | - | 2 | 778 | 30322852 |