Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC367195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047920.1 | NP_001041385.1 | 2 | 152 | ||
GenBank mRNA | AY325186.1 | - | 2 | 152 |