Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012225.2 | NP_001012225.1 | 11, 12 | 1413 | ||
NM_001160155.1 | NP_001153627.1 | 11, 12 | 1440 | |||
XM_006244840.2 | XP_006244902.1 | 11, 12 | 1465 | |||
XM_008767040.1 | XP_008765262.1 | 10, 11 | 1128 | |||
GenBank mRNA | BC088215.1 | - | 10, 11 | 1371 | 7383944 |