Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109006.1 | NP_001102476.1 | 9 | 1289 | ||
XM_006245260.2 | XP_006245322.1 | 10 | 1575 | |||
XM_008767280.1 | XP_008765502.1 | 9 | 1468 | |||
XM_008767281.1 | XP_008765503.1 | 10 | 1296 | |||
XM_008767282.1 | XP_008765504.1 | 9 | 1062 |