Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Praf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109013.1 | NP_001102483.1 | 1 | 288 | ||
GenBank mRNA | FQ213026.1 | - | 1 | 147 | ||
FQ213646.1 | - | 1 | 144 | |||
FQ219908.1 | - | 1 | 147 |