Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107442.1 | NP_001100912.1 | 5 | 1553 | ||
XM_006255812.2 | XP_006255874.1 | 4 | 2621 | |||
XM_008772614.1 | XP_008770836.1 | 4 | 1441 | |||
XM_008772615.1 | XP_008770837.1 | 4 | 1617 | |||
GenBank mRNA | BC098861.1 | - | 1 | 56 | ||
BC160901.1 | - | 4 | 1455 | 7454090 |
Gene Symbol: LOC690283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758820.1 | XP_008757042.1 | 1 | 1637 |