Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031099.1 | NP_112361.1 | 5 | 675 | ||
XM_006250585.2 | XP_006250647.1 | 5 | 656 | |||
GenBank mRNA | BC060561.1 | - | 5 | 622 | 6918805 | |
FQ212710.1 | - | 5 | 632 | |||
FQ214493.1 | - | 5 | 623 | |||
FQ215029.1 | - | 5 | 624 | |||
FQ219244.1 | - | 5 | 621 | |||
FQ219787.1 | - | 5 | 623 | |||
FQ219848.1 | - | 5 | 623 | |||
FQ220068.1 | - | 5 | 623 | |||
FQ220161.1 | - | 5 | 623 | |||
FQ220170.1 | - | 5 | 623 | |||
FQ220205.1 | - | 5 | 623 | |||
FQ220219.1 | - | 5 | 623 | |||
FQ227439.1 | - | 5 | 623 | |||
FQ228473.1 | - | 5 | 623 | |||
FQ229451.1 | - | 5 | 624 | |||
FQ229816.1 | - | 5 | 621 | |||
FQ229964.1 | - | 5 | 625 | |||
FQ229979.1 | - | 5 | 623 | |||
FQ230017.1 | - | 5 | 621 | |||
FQ230061.1 | - | 5 | 626 | |||
FQ230102.1 | - | 5 | 623 | |||
FQ230128.1 | - | 5 | 651 | |||
FQ230161.1 | - | 5 | 649 | |||
FQ230794.1 | - | 5 | 626 | |||
FQ231729.1 | - | 5 | 621 | |||
FQ232540.1 | - | 5 | 623 | |||
FQ233314.1 | - | 5 | 648 | |||
FQ234171.1 | - | 5 | 653 | |||
M17419.1 | - | 5 | 608 | |||
X06148.1 | - | 5 | 675 |