Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135110.2 | NP_001128582.1 | 6 | 990 | ||
NM_001286516.1 | NP_001273445.1 | 4 | 641 | |||
NM_005147.5 | NP_005138.3 | 6 | 990 | |||
GenBank mRNA | AB209923.1 | - | 6 | 909 | ||
AF061749.1 | - | 6 | 944 | |||
AF244136.1 | - | 6 | 915 | |||
AF411044.1 | - | 6 | 957 | |||
AK001333.1 | - | 6 | 930 | |||
AK223105.1 | - | 6 | 930 | |||
AK295391.1 | - | 4 | 581 | |||
AK314218.1 | - | 6 | 927 | |||
BC007225.1 | - | 6 | 910 | |||
BC011855.2 | - | 6 | 914 | 4139881 | ||
BC012343.1 | - | 6 | 906 | 4653507 | ||
BC014062.2 | - | 6 | 906 | |||
BC020248.1 | - | 2 | 239 | |||
BC030145.2 | - | 6 | 906 | |||
BC032100.2 | - | 6 | 928 | 4660602 |