Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135110.2 | NP_001128582.1 | 9 | 1297 | ||
NM_001286516.1 | NP_001273445.1 | 7 | 948 | |||
NM_005147.5 | NP_005138.3 | 9 | 1297 | |||
GenBank mRNA | AB209923.1 | - | 9 | 1216 | ||
AF061749.1 | - | 9 | 1251 | |||
AF411044.1 | - | 9 | 1264 | |||
AK001333.1 | - | 9 | 1237 | |||
AK223105.1 | - | 9 | 1237 | |||
AK295391.1 | - | 7 | 888 | |||
AK314218.1 | - | 9 | 1234 | |||
BC007225.1 | - | 9 | 1217 | |||
BC011855.2 | - | 9 | 1221 | 4139881 | ||
BC012343.1 | - | 9 | 1213 | 4653507 | ||
BC014062.2 | - | 9 | 1213 | |||
BC020248.1 | - | 5 | 546 | |||
BC030145.2 | - | 9 | 1213 | |||
BC032100.2 | - | 9 | 1235 | 4660602 |