Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257709.2 | XP_006257771.1 | 2 | 6467 | ||
XM_006257710.2 | XP_006257772.1 | 2 | 684 | |||
XM_008773714.1 | XP_008771936.1 | 3 | 1016 |
Gene Symbol: Hoxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762948.1 | XP_008761170.1 | 2 | 6467 | ||
XM_008762949.1 | XP_008761171.1 | 3 | 1016 | |||
XM_008762950.1 | XP_008761172.1 | 2 | 684 |
Gene Symbol: LOC100912608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236522.2 | XP_006236584.1 | 684 | |||
XM_006236523.2 | XP_006236585.1 | 6479 | ||||
XM_006236524.2 | XP_006236586.1 | 6392 |