Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myadm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183332.1 | NP_899161.1 | 3 | 1014 | ||
XM_006228035.2 | XP_006228097.1 | 3 | 1205 | |||
GenBank mRNA | AY344060.1 | - | 3 | 1014 | ||
BC087054.1 | - | 2 | 925 | 7123822 | ||
BC107558.1 | - | 2 | 1026 | 7932126 | ||
BC128719.1 | - | 3 | 1171 | 7315920 |