Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199509.1 | NP_955803.1 | 1 | 2392 | ||
GenBank mRNA | AB113397.1 | - | 1 | 2392 | ||
AY573984.1 | - | 1 | 2392 |