Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243759.1 | NP_001230688.1 | 8 | 861 | ||
NM_004205.4 | NP_004196.4 | 9 | 1676 | |||
NM_171997.2 | NP_741994.1 | 8 | 941 | |||
XM_005271721.4 | XP_005271778.1 | 9 | 1549 | |||
XM_005271722.2 | XP_005271779.1 | 9 | 1555 | |||
XM_017018539.1 | XP_016874028.1 | 8 | 637 | |||
GenBank mRNA | AF079564.1 | - | 8 | 679 | ||
AF440755.1 | - | 8 | 941 | |||
AK057225.1 | - | 9 | 1605 | |||
AK292255.1 | - | 8 | 864 | |||
AK301805.1 | - | 8 | 746 | |||
BC002854.1 | - | 9 | 1557 | 3635143 | ||
BC002955.1 | - | 9 | 1557 | 3543435 | ||
BC041366.2 | - | 8 | 846 | 5273400 |