Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000601.1 | NP_001000601.1 | 1 | 763 |
Gene Symbol: Olr783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000376.1 | NP_001000376.1 | 1 | 763 |