Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000606.1 | NP_001000606.1 | 1 | 61 |
Gene Symbol: Olr779 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000917.1 | NP_001000917.1 | 1 | 61 |