Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128610.2 | NP_001122082.1 | 14 | 2561 | ||
NM_001283049.1 | NP_001269978.1 | 11 | 2103 | |||
NM_005154.4 | NP_005145.3 | 14 | 2421 | |||
XM_006720761.3 | XP_006720824.1 | 15 | 2741 | |||
XM_006720762.3 | XP_006720825.1 | 14 | 2654 | |||
XM_011522193.2 | XP_011520495.1 | 15 | 2578 | |||
XM_017022718.1 | XP_016878207.1 | 13 | 2329 | |||
XM_017022719.1 | XP_016878208.1 | 13 | 2446 | |||
XM_017022720.1 | XP_016878209.1 | 14 | 2491 | |||
XM_017022721.1 | XP_016878210.1 | 15 | 2478 | |||
XM_017022722.1 | XP_016878211.1 | 13 | 2401 | |||
GenBank mRNA | AK292400.1 | - | 14 | 2416 | ||
AK296480.1 | - | 11 | 2093 | |||
AK300590.1 | - | 3 | 652 | |||
BC110590.2 | - | 14 | 2387 | 40027270 | ||
BX537420.1 | - | 14 | 2416 | |||
D29956.1 | - | 14 | 2535 |