Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282848.1 | NP_001269777.1 | 6 | 1518 | ||
NM_001282849.1 | NP_001269778.1 | 5 | 1351 | |||
NM_004683.5 | NP_004674.1 | 5 | 947 | |||
NM_152869.3 | NP_690608.1 | 6 | 1567 | |||
XM_006724567.2 | XP_006724630.1 | 4 | 667 | |||
GenBank mRNA | AB028125.1 | - | 5 | 671 | ||
AB032064.1 | - | 5 | 729 | |||
AK223360.1 | - | 5 | 947 | |||
AK290136.1 | - | 5 | 885 | |||
BC050371.1 | - | 5 | 1289 | 5760604 | ||
BC073173.1 | - | 5 | 761 | 6186669 | ||
D31815.1 | - | 5 | 670 | |||
EU794655.1 | - | 5 | 729 |