Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000646.1 | NP_001000646.1 | 1 | 625 |
Gene Symbol: LOC100910893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234497.2 | XP_006234559.1 | 622 |
Gene Symbol: LOC103690355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762002.1 | XP_008760224.1 | 1 | 622 |
Gene Symbol: Olr637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001126.1 | NP_001001126.1 | 625 |