Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004687.4 | NP_004678.3 | 13 | 1450 | ||
XM_005257784.2 | XP_005257841.1 | 12 | 1599 | |||
XM_005257785.4 | XP_005257842.1 | 14 | 1567 | |||
XM_005257786.4 | XP_005257843.1 | 13 | 2354 | |||
XM_006722168.3 | XP_006722231.1 | 13 | 1394 | |||
XM_011525460.2 | XP_011523762.1 | 14 | 2447 | |||
GenBank mRNA | AB014547.2 | - | 11 | 1340 | ||
AF264717.1 | - | 13 | 1393 | |||
BC035609.1 | - | 13 | 1411 | 5201989 |