Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000664.1 | NP_001000664.1 | 1 | 58 |
Gene Symbol: Olr575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000327.1 | NP_001000327.1 | 1 | 58 |