Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004691.4 | NP_004682.2 | 8 | 1091 | ||
XM_011523444.1 | XP_011521746.1 | 9 | 1078 | |||
XM_011523445.1 | XP_011521747.1 | 9 | 1211 | |||
XM_011523446.1 | XP_011521748.1 | 8 | 983 | |||
GenBank mRNA | AK128641.1 | - | 4 | 2811 | ||
AK300570.1 | - | 9 | 1146 | |||
AK301630.1 | - | 9 | 1080 | |||
AK313035.1 | - | 8 | 1049 | |||
BC008861.2 | - | 8 | 1069 | 4126712 | ||
L05087.1 | - | 3 | 202 | |||
X71490.1 | - | 8 | 1016 |