Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032727.3 | NP_116116.1 | 3 | 1379 | ||
GenBank mRNA | AB209785.1 | - | 3 | 830 | ||
AK293490.1 | - | 3 | 393 | |||
BC006359.2 | - | 3 | 1379 | 4125949 | ||
S78296.1 | - | 3 | 1360 |