Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174166.1 | NP_001167637.1 | 7 | 1815 | ||
NM_004694.4 | NP_004685.2 | 6 | 1688 | |||
XM_005257789.3 | XP_005257846.2 | 6 | 1853 | |||
XM_011525461.2 | XP_011523763.1 | 6 | 1532 | |||
XM_017025291.1 | XP_016880780.1 | 7 | 1652 | |||
XM_017025292.1 | XP_016880781.1 | 6 | 1715 | |||
GenBank mRNA | AK290417.1 | - | 6 | 1564 | ||
AK313121.1 | - | 6 | 1554 | |||
AX750581.1 | - | 7 | 1667 | |||
AX750582.1 | - | 5 | 1502 | |||
BC064832.1 | - | 6 | 1690 | 6498761 | ||
BC127749.1 | - | 2 | 552 | |||
BC127750.1 | - | 2 | 550 | |||
BC141962.1 | - | 2 | 551 | |||
BC150547.1 | - | 2 | 552 | |||
BC150548.1 | - | 2 | 552 | |||
U79745.1 | - | 7 | 1667 |