Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000746.1 | NP_001000746.1 | 1 | 442 |
Gene Symbol: Olr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000745.1 | NP_001000745.1 | 1 | 445 |