Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174166.1 | NP_001167637.1 | 6 | 1277 | ||
NM_004694.4 | NP_004685.2 | 5 | 1150 | |||
XM_005257789.3 | XP_005257846.2 | 5 | 1315 | |||
XM_011525461.2 | XP_011523763.1 | 5 | 994 | |||
XM_017025291.1 | XP_016880780.1 | 6 | 1114 | |||
XM_017025292.1 | XP_016880781.1 | 5 | 1177 | |||
GenBank mRNA | AK290417.1 | - | 5 | 1026 | ||
AK313121.1 | - | 5 | 1016 | |||
AX750581.1 | - | 6 | 1129 | |||
AX750582.1 | - | 4 | 964 | |||
BC064832.1 | - | 5 | 1152 | 6498761 | ||
BC127749.1 | - | 1 | 14 | |||
BC127750.1 | - | 1 | 12 | |||
BC141962.1 | - | 1 | 13 | |||
BC150547.1 | - | 1 | 14 | |||
BC150548.1 | - | 1 | 14 | |||
U79745.1 | - | 6 | 1129 |