Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000762.1 | NP_001000762.1 | 1 | 469 | ||
XM_008760258.1 | XP_008758480.1 | 3 | 1023 | |||
XM_008760259.1 | XP_008758481.1 | 4 | 918 |
Gene Symbol: Olr323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000245.1 | NP_001000245.1 | 1 | 469 |