Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000769.1 | NP_001000769.1 | 1 | 847 |
Gene Symbol: Olr1457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000526.1 | NP_001000526.1 | 1 | 847 |
Gene Symbol: Olr1458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000021.1 | NP_001000021.1 | 1 | 847 |
Gene Symbol: Olr1459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000770.1 | NP_001000770.1 | 1 | 847 |