Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000790.1 | NP_001000790.1 | 476 | |||
XM_008766124.1 | XP_008764346.1 | 2 | 365 |
Gene Symbol: Olr1326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000474.1 | NP_001000474.1 | 1 | 476 |
Gene Symbol: Olr1332 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000477.1 | NP_001000477.1 | 1 | 476 |