Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000800.1 | NP_001000800.1 | 448 |
Gene Symbol: LOC100910010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749260.3 | XP_003749308.3 | 1 | 448 |
Gene Symbol: LOC100910041 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749182.3 | XP_003749230.3 | 1 | 448 |
Gene Symbol: LOC100910256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750492.2 | XP_003750540.2 | 448 |
Gene Symbol: LOC100910335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750494.2 | XP_003750542.2 | 1 | 448 |
Gene Symbol: LOC100910445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750496.3 | XP_003750544.3 | 1 | 448 |