Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000814.1 | NP_001000814.1 | 1 | 672 |
Gene Symbol: LOC100912317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242838.2 | XP_006242900.1 | 672 |
Gene Symbol: LOC102554388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242834.1 | XP_006242896.1 | 1 | 672 |