Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000839.1 | NP_001000839.1 | 1 | 836 |
Gene Symbol: Olr1616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000840.1 | NP_001000840.1 | 1 | 836 |
Set Membership: |
G-protein coupled receptor |