Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130846.3 | NP_001124318.2 | 4 | 1214 | ||
NM_001130847.3 | NP_001124319.1 | 14 | 1760 | |||
NM_004208.3 | NP_004199.1 | 13 | 1557 | |||
NM_145812.2 | NP_665811.1 | 13 | 1545 | |||
NR_132647.1 | 13 | 1617 | ||||
GenBank mRNA | AF100928.1 | - | 13 | 1368 | ||
AF131759.1 | - | 10 | 909 | |||
AK000775.1 | - | 13 | 1393 | |||
AK314446.1 | - | 13 | 1345 | |||
AL049703.1 | - | 13 | 1410 | |||
AL049704.1 | - | 5 | 489 | |||
BC111065.1 | - | 13 | 1488 | 5767978 | ||
BC139738.1 | - | 13 | 1461 | 40085139 | ||
CR457379.1 | - | 13 | 1326 | |||
DQ016496.2 | - | 4 | 1214 | |||
DQ016498.1 | - | 14 | 1537 | |||
DQ016500.1 | - | 13 | 1394 |