Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000865.1 | NP_001000865.1 | 1 | 209 |
Gene Symbol: Olr1166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000867.1 | NP_001000867.1 | 1 | 209 |