Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000878.1 | NP_001000878.1 | 1 | 244 |
Gene Symbol: LOC103693022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766028.1 | XP_008764250.1 | 1 | 244 |
Gene Symbol: Olr1117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000884.1 | NP_001000884.1 | 1 | 244 |