Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057749.2 | NP_477097.1 | 5 | 325 | ||
XM_011517366.2 | XP_011515668.1 | 4 | 650 | |||
XM_017013957.1 | XP_016869446.1 | 4 | 650 | |||
XM_017013958.1 | XP_016869447.1 | 5 | 467 | |||
XM_017013959.1 | XP_016869448.1 | 5 | 569 | |||
GenBank mRNA | AF091433.1 | - | 4 | 258 | ||
AF102778.1 | - | 3 | 221 | |||
AF106690.1 | - | 3 | 221 | |||
AF112857.1 | - | 3 | 221 | |||
BC020729.1 | - | 5 | 272 | 4774554 | ||
BG720611.1 | - | 5 | 330 | |||
BJ990268.1 | - | 5 | 325 |