Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000895.1 | NP_001000895.1 | 1 | 794 |
Gene Symbol: LOC100912354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751901.3 | XP_003751949.3 | 1 | 395 |
Gene Symbol: Olr1679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000892.1 | NP_001000892.1 | 794 |
Gene Symbol: Olr1680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000891.1 | NP_001000891.1 | 1 | 794 |
Gene Symbol: Olr1681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001006.1 | NP_001001006.1 | 1 | 794 |
Gene Symbol: Olr1682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000890.1 | NP_001000890.1 | 1 | 794 |
Gene Symbol: Olr1683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000269.1 | NP_001000269.1 | 1 | 794 |