Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083585.2 | NP_001077054.1 | 10 | 1837 | ||
NM_001291581.1 | NP_001278510.1 | 9 | 1708 | |||
NM_004703.5 | NP_004694.2 | 10 | 1837 | |||
XM_011524060.1 | XP_011522362.1 | 11 | 1903 | |||
XM_017025294.1 | XP_016880783.1 | 9 | 1430 | |||
XM_017025295.1 | XP_016880784.1 | 8 | 1364 | |||
GenBank mRNA | AF098638.1 | - | 10 | 1613 | ||
AK297535.1 | - | 9 | 1697 | |||
AK310932.1 | - | 9 | 1649 | |||
AK314183.1 | - | 10 | 1828 | |||
BC032435.1 | - | 10 | 1791 | |||
BC041700.1 | - | 10 | 1803 | 6046320 | ||
X77723.1 | - | 10 | 1686 | |||
X91141.1 | - | 10 | 1801 |