Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004706.3 | NP_004697.2 | 8 | 737 | ||
NM_198977.1 | NP_945328.1 | 7 | 638 | |||
NM_199002.1 | NP_945353.1 | 8 | 762 | |||
XM_005259386.4 | XP_005259443.1 | 8 | 1089 | |||
XM_005259390.4 | XP_005259447.1 | 8 | 1085 | |||
XM_011527468.2 | XP_011525770.1 | 8 | 1085 | |||
GenBank mRNA | BC005155.2 | - | 5 | 475 | ||
BC011726.2 | - | 7 | 613 | 4138058 | ||
BC034013.1 | - | 8 | 693 | 3451036 | ||
BC067262.1 | - | 7 | 581 | 6502165 | ||
BT007421.1 | - | 5 | 451 | |||
U64105.1 | - | 8 | 682 |